Learn More
New England Biolabs, Inc. Monarch® DNA/RNA Protection Reagent - 56 ml
SDP
Supplier:  New England Biolabs, Inc. T2011L
Encompass
Monarch DNA/RNA Protection Reagent is a component of the Monarch Total RNA Miniprep Kit. It is a solution for the preservation of nucleic acids in various tissue types and is supplied as a 2X concentrate. This reagent does not need to be removed before RNA extraction with the Monarch Total RNA Miniprep Kit.
  • Preserves and stabilizes RNA and DNA
  • A component of the Monarch Total RNA Miniprep Kit
  • Supplied as a 2X concentrateSupplied in 2 bottles, each containing 28 ml
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-152-7887
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product