Learn More
New England Biolabs, Inc. Monarch DNase I Lyo 2200U
SDP
Catalog No. 5000077840
Encompass
Monarch DNase I Lyophilized is a lyophilized format of DNA specific Endonuclease DNase I supplied in a dry cake It is also included as a component of the Monarch Spin RNA Isolation Kit After rehydration with nuclease-free water the resulting enzyme solution is suitable for preparing DNase reactions with the supplied reaction buffer Optimal DNA removal is achieved either on-column or in-solution DNase I acts on single- and double-stranded DNA chromatin and RNA DNA hybrids
Catalog No. 50-000-77840 Supplier New England Biolabs, Inc. Supplier No. T2104S
Monarch DNase I Lyophilized is a lyophilized format of DNA specific Endonuclease DNase I supplied in a dry cake It is also included as a component of the Monarch Spin RNA Isolation Kit After rehydration with nuclease-free water the resulting enzyme solution is suitable for preparing DNase reactions with the supplied reaction buffer Optimal DNA removal is achieved either on-column or in-solution DNase I acts on single- and double-stranded DNA chromatin and RNA DNA hybrids