Learn More
New England Biolabs, Inc. Monarch gDNA Elution Buffer II - 24 ml
SDP
Catalog No. 502068311
Encompass
The Monarch gDNA Elution Buffer II is supplied as a component of the Monarch HMW DNA Extraction Kits. It is formulated for long term storage of genomic DNA and is made of 10mM Tris, pH 9.0, 0.5 mM EDTA. This product is supplied as 2 bottles of 12 ml.
  • Supplied as a component of the Monarch HMW DNA Extraction Kits
  • Optimized for long term storage of HMW gDNA
  • 10mM Tris, pH 9.0, 0.5 mM EDTA
Catalog No. 50-206-8311 Supplier New England Biolabs, Inc. Supplier No. T3056L
The Monarch gDNA Elution Buffer II is supplied as a component of the Monarch HMW DNA Extraction Kits. It is formulated for long term storage of genomic DNA and is made of 10mM Tris, pH 9.0, 0.5 mM EDTA. This product is supplied as 2 bottles of 12 ml.
  • Supplied as a component of the Monarch HMW DNA Extraction Kits
  • Optimized for long term storage of HMW gDNA
  • 10mM Tris, pH 9.0, 0.5 mM EDTA