Learn More
New England Biolabs, Inc. Monarch® gDNA Nuclei Prep & Lysis Buffer Pack - 1 pack
SDP
Catalog No. 502068309
Encompass
The Monarch gDNA Nuclei Prep & Lysis Buffer Pack contains two lysis buffers used in the Monarch HMW DNA Extraction Kit for Cells & Blood. The Nuclei Prep Buffer is a mild lysis buffer used first to lyse the cell membrane and release the contents of the cytoplasm while the nuclei are kept intact. It is used pre-mixed with the RNase A enzyme included in the kit. After the RNA from the cytoplasm is digested by RNase A, the Nuclei Lysis Buffer, pre-mixed with the included Proteinase K, is used to lyse the nuclei and release the HMW genomic DNA. This 2-step lysis ensures that RNA in the cytoplasm is effectively degraded before the viscous genomic DNA from the nuclei is released. 20 ml of each buffer is included in this buffer pack. The Nuclei Prep Buffer is used chilled to 4°C, and it can be stored at 4°C for convenience if desired.
  • Supplied as components of the Monarch HMW DNA Extraction Kit for Cells & Blood
Catalog No. 50-206-8309 Supplier New England Biolabs, Inc. Supplier No. T3054L
The Monarch gDNA Nuclei Prep & Lysis Buffer Pack contains two lysis buffers used in the Monarch HMW DNA Extraction Kit for Cells & Blood. The Nuclei Prep Buffer is a mild lysis buffer used first to lyse the cell membrane and release the contents of the cytoplasm while the nuclei are kept intact. It is used pre-mixed with the RNase A enzyme included in the kit. After the RNA from the cytoplasm is digested by RNase A, the Nuclei Lysis Buffer, pre-mixed with the included Proteinase K, is used to lyse the nuclei and release the HMW genomic DNA. This 2-step lysis ensures that RNA in the cytoplasm is effectively degraded before the viscous genomic DNA from the nuclei is released. 20 ml of each buffer is included in this buffer pack. The Nuclei Prep Buffer is used chilled to 4°C, and it can be stored at 4°C for convenience if desired.
  • Supplied as components of the Monarch HMW DNA Extraction Kit for Cells & Blood