Learn More
New England Biolabs, Inc. Monarch Spin DNA Gel Extraction Kit - 50 preps
SDP
Catalog No. 502722965
Encompass
The Monarch Spin DNA Gel Extraction Kit, a newer version of our original gel extraction kit, offers an efficient method for DNA gel extraction and purification, enabling up to 5 ug of high-quality DNA. This new version features upgraded spin columns precision-engineered to uniquely allow for low elution, in as little as 5 ul, for highly concentrated DNA in downstream applications such as sequencing, restriction digestion, library construction, labeling and other enzymatic manipulations. Our optimized gel dissolving buffer allows efficient gel dissolving and DNA binding on the column, without a need to monitor pH or add isopropanol. These kits contain uniquely designed columns and bottles to significantly reduce plastic usage compared to leading suppliers, while maintaining structural integrity for centrifugation, vacuum manifolds and other steps
Catalog No. 50-272-2965 Supplier New England Biolabs, Inc. Supplier No. T1120S
The Monarch Spin DNA Gel Extraction Kit, a newer version of our original gel extraction kit, offers an efficient method for DNA gel extraction and purification, enabling up to 5 ug of high-quality DNA. This new version features upgraded spin columns precision-engineered to uniquely allow for low elution, in as little as 5 ul, for highly concentrated DNA in downstream applications such as sequencing, restriction digestion, library construction, labeling and other enzymatic manipulations. Our optimized gel dissolving buffer allows efficient gel dissolving and DNA binding on the column, without a need to monitor pH or add isopropanol. These kits contain uniquely designed columns and bottles to significantly reduce plastic usage compared to leading suppliers, while maintaining structural integrity for centrifugation, vacuum manifolds and other steps