Learn More
Enterprise Technology Solutions Monarch Global Brands Cloths,Microfiber,Midweight,16"x16",50/BX,YW
SDP
Catalog No. 5000790342
Encompass_Preferred
Microfiber cloths are cost-effectively engineered for your convenience. Reusable design lets you launder hundreds of times. Large size makes it easier to use by hand. Precision-cut, seamless edges efficiently clean either wet or dry while removing heavy soils and grime for a streak-free, lint-free finish. Mid-weight cloths are ideal for utility carts, industrial/automotive shops, offices, supply rooms, healthcare facilities and households. Center-pull dispenser box allows you to grab the rag you need with ease.
Catalog No. 50-007-90342 Supplier Enterprise Technology Solutions Supplier No. MONM931Y
Microfiber cloths are cost-effectively engineered for your convenience. Reusable design lets you launder hundreds of times. Large size makes it easier to use by hand. Precision-cut, seamless edges efficiently clean either wet or dry while removing heavy soils and grime for a streak-free, lint-free finish. Mid-weight cloths are ideal for utility carts, industrial/automotive shops, offices, supply rooms, healthcare facilities and households. Center-pull dispenser box allows you to grab the rag you need with ease.