Learn More
New England Biolabs, Inc. Monarch® HMW DNA Extraction Kit for Tissue - 50 preps
SDP
Catalog No. 502068302
Encompass
The Monarch HMW DNA Extraction Kit for Tissue provides a rapid and reliable process for extracting high molecular weight (HMW), intact genomic DNA from various tissues and bacteria, as well as other sample types including yeast, insect, and amphibian. Optimized extraction protocol for tissue utilizes pestle homogenization and proteinase K digestion with agitation for sample lysis, followed by a protein removal step and precipitation of the extracted DNA onto the surface of large glass beads. DNA size ranges from 50 to greater than or equal to 500 kb for standard protocols and into the Mb range when the lowest agitation speeds are used for processing soft organ tissues and bacteria. Purified DNA is recovered in high yield with excellent purity, including nearly complete removal of RNA. Purified HMW DNA is suitable for a variety of downstream applications including long-read sequencing
Catalog No. 50-206-8302 Supplier New England Biolabs, Inc. Supplier No. T3060L
The Monarch HMW DNA Extraction Kit for Tissue provides a rapid and reliable process for extracting high molecular weight (HMW), intact genomic DNA from various tissues and bacteria, as well as other sample types including yeast, insect, and amphibian. Optimized extraction protocol for tissue utilizes pestle homogenization and proteinase K digestion with agitation for sample lysis, followed by a protein removal step and precipitation of the extracted DNA onto the surface of large glass beads. DNA size ranges from 50 to greater than or equal to 500 kb for standard protocols and into the Mb range when the lowest agitation speeds are used for processing soft organ tissues and bacteria. Purified DNA is recovered in high yield with excellent purity, including nearly complete removal of RNA. Purified HMW DNA is suitable for a variety of downstream applications including long-read sequencing