Learn More
MSC Monarch Labels, Blue, 3/4 x 5/8 in, Permanent, 8 Rolls/Box
Catalog No. 5000789097
Encompass
These blue permanent labels are designed for quick and accurate marking of price, date, or inventory. Each box includes 8 rolls, totaling 14,000 labels, and a free ink roller.
  • Suitable for use with Monarch 1136 Numeric and Alpha/Numeric label guns
  • Label size of 3/4 x 5/8 in
  • Each roll contains 1,750 labels
  • Permanent adhesive for secure application
  • No security slits for continuous use
  • Can be used in freezer conditions down to -10°F
  • Shelf life of 1 year
Catalog No. 50-007-89097 Supplier MSC Supplier No. 55600563
These blue permanent labels are designed for quick and accurate marking of price, date, or inventory. Each box includes 8 rolls, totaling 14,000 labels, and a free ink roller.
  • Suitable for use with Monarch 1136 Numeric and Alpha/Numeric label guns
  • Label size of 3/4 x 5/8 in
  • Each roll contains 1,750 labels
  • Permanent adhesive for secure application
  • No security slits for continuous use
  • Can be used in freezer conditions down to -10°F
  • Shelf life of 1 year