Learn More
New England Biolabs, Inc. Monarch Mag PCR & DNA Cleanup Kit (5 ug) - 10 preps
SDP
Supplier:  New England Biolabs, Inc. T4130V
Encompass
The Monarch Mag PCR & DNA Cleanup Kit (5 ug) enables rapid and reliable purification and concentration of up to 5 μg of DNA, including both single- and double-stranded forms, from common enzymatic reactions such as PCR, restriction digestion, ligation, reverse transcription, and rolling circle amplification (RCA). Using a streamlined bind-wash-elute workflow with silica-coated magnetic beads, the kit delivers highly pure, inhibitor-free DNA ready for immediate use in downstream applications such as in vitro transcription (IVT), sequencing, restriction digestion, library construction, labeling, and other enzymatic manipulations. The magnetic bead format offers flexibility and scalability, supporting both manual and automated workflows. Additional modified protocols support varying DNA input amounts and sample volumes and enable purification of oligos, gDNA, and RCA products. The kit includes magnetic beads, optimized buffers, and reagents, all compactly packaged.
Catalog No. 50-003-32606