Learn More
New England Biolabs, Inc. Monarch® Mag Viral DNA/RNA Extraction Kit - 3X 600 preps
SDP
Catalog No. 502581798
Encompass
The Monarch Mag Viral DNA/RNA Extraction Kit is designed for hands-free isolation of viral nucleic acid from saliva, respiratory-swab samples using automated processing and liquid handling systems. The kit features a magnetic bead-based method for extraction and purification and is compatible with automated instruments such as KingFisher, Agilent Bravo, and more, as well as manual workflows. The kit enables highly sensitive detection and amplification in qPCR/RT-qPCR, ddPCR applications and is compatible with other downstream applications such as sequencing/NGS. Carrier RNA is included as a co-precipitant in the workflow to enhance the recovery of low amounts of viral RNA.
  • Designed for hands-free extraction of viral DNA and/or RNA
  • Utilizes magnetic bead-based methods, supporting automated and manual workflows
  • Developed for highly sensitive amplification, sequencing, and other downstream applications
Catalog No. 50-258-1798 Supplier New England Biolabs, Inc. Supplier No. T4010X
The Monarch Mag Viral DNA/RNA Extraction Kit is designed for hands-free isolation of viral nucleic acid from saliva, respiratory-swab samples using automated processing and liquid handling systems. The kit features a magnetic bead-based method for extraction and purification and is compatible with automated instruments such as KingFisher, Agilent Bravo, and more, as well as manual workflows. The kit enables highly sensitive detection and amplification in qPCR/RT-qPCR, ddPCR applications and is compatible with other downstream applications such as sequencing/NGS. Carrier RNA is included as a co-precipitant in the workflow to enhance the recovery of low amounts of viral RNA.
  • Designed for hands-free extraction of viral DNA and/or RNA
  • Utilizes magnetic bead-based methods, supporting automated and manual workflows
  • Developed for highly sensitive amplification, sequencing, and other downstream applications