Learn More
New England Biolabs, Inc. Monarch PCR & DNA Cleanup Kit (5 µg) – 50 preps
SDP
Catalog No. NC1210607
Encompass
The Monarch PCR and DNA Cleanup Kit rapidly and reliably purifies up to 5 ug of concentrated, high-quality DNA from PCR and other enzymatic reactions. The kit utilizes a bind/wash/elute workflow with minimal incubation and spin times. The columns ensure zero buffer retention and no carryover of contaminants, enabling elution of sample in volumes as low as 6 ul. The buffers provided have been optimized, and do not require monitoring of pH. Eluted DNA is ready for use in restriction digests, DNA sequencing, ligation and other enzymatic manipulations. Designed with sustainability in mind, Monarch kits use significantly less plastic and responsibly-sourced, recyclable packaging. The protocol can also be modified to enable the purification of smaller DNA fragments, including oligonucleotides and ssDNA.
Catalog No. NC1210607 Supplier New England Biolabs, Inc. Supplier No. T1030S
The Monarch PCR and DNA Cleanup Kit rapidly and reliably purifies up to 5 ug of concentrated, high-quality DNA from PCR and other enzymatic reactions. The kit utilizes a bind/wash/elute workflow with minimal incubation and spin times. The columns ensure zero buffer retention and no carryover of contaminants, enabling elution of sample in volumes as low as 6 ul. The buffers provided have been optimized, and do not require monitoring of pH. Eluted DNA is ready for use in restriction digests, DNA sequencing, ligation and other enzymatic manipulations. Designed with sustainability in mind, Monarch kits use significantly less plastic and responsibly-sourced, recyclable packaging. The protocol can also be modified to enable the purification of smaller DNA fragments, including oligonucleotides and ssDNA.