Learn More
New England Biolabs, Inc. Monarch® Pestle Set - 100 sets
SDP
Catalog No. 502068305
Encompass
The Monarch Pestle Set contains single-use microtube pestles and the accompanying 1.5 ml tubes for sample disruption and homogenization prior to nucleic acid extraction. 100 green pestles and 100 tubes are included in this set.
  • Set includes Monarch Pestles and Monarch Pestle Tubes
  • Optimally designed for manual homogenization of tissue samples prior to nucleic acid extraction
  • Single-use pestles and tubes
Catalog No. 50-206-8305 Supplier New England Biolabs, Inc. Supplier No. T3000L
The Monarch Pestle Set contains single-use microtube pestles and the accompanying 1.5 ml tubes for sample disruption and homogenization prior to nucleic acid extraction. 100 green pestles and 100 tubes are included in this set.
  • Set includes Monarch Pestles and Monarch Pestle Tubes
  • Optimally designed for manual homogenization of tissue samples prior to nucleic acid extraction
  • Single-use pestles and tubes