Learn More
New England Biolabs, Inc. Monarch® Precipitation Enhancer - 10 ml
SDP
Catalog No. 502783701
Encompass
The Monarch Precipitation Enhancer Solution, supplied as a component of the Monarch HMW DNA Extraction Kit for Cells & Blood, is a salt-based solution that facilitates the precipitation of DNA onto the Monarch DNA Capture Beads during HMW DNA extraction from cells and blood. It is supplied in 2 bottles of 5 ml.
  • A component of the Monarch HMW DNA Extraction Kit for Cells & Blood
  • Salt-based solution using in the HMW DNA extraction workflow
  • Facilitates DNA precipitation onto the glass beads during HMW DNA extraction from cells & blood
  • Supplied as 2 bottles of 5 ml
Catalog No. 50-278-3701 Supplier New England Biolabs, Inc. Supplier No. T3055L
The Monarch Precipitation Enhancer Solution, supplied as a component of the Monarch HMW DNA Extraction Kit for Cells & Blood, is a salt-based solution that facilitates the precipitation of DNA onto the Monarch DNA Capture Beads during HMW DNA extraction from cells and blood. It is supplied in 2 bottles of 5 ml.
  • A component of the Monarch HMW DNA Extraction Kit for Cells & Blood
  • Salt-based solution using in the HMW DNA extraction workflow
  • Facilitates DNA precipitation onto the glass beads during HMW DNA extraction from cells & blood
  • Supplied as 2 bottles of 5 ml