Learn More
New England Biolabs, Inc. Monarch RBC Lysis Buffer - 160 ml
SDP
Catalog No. 502068310
Encompass
Supplied as a component of the Monarch HMW DNA Extraction Kit for Cells & Blood, the Monarch RBC Lysis buffer is specifically designed to lyse red blood cells (erythrocytes) while leaving leukocytes intact. The separate RBC lysis step enables removal of hemoglobin to increase the purity of the blood prep. The RBC Lysis Buffer also removes nucleases to enable the subsequent purification of HMW DNA into the megabase (Mb) range in size. This product should be stored at 4°C.
  • A component of the Monarch HMW DNA Extraction Kit for Cells & Blood
  • Lyses red blood cells (erythrocytes) while leaving leukocytes intact
  • Enables removal of hemoglobin and nucleases in blood preps
  • Store Monarch RBC Lysis Buffer at 4°C
Catalog No. 50-206-8310 Supplier New England Biolabs, Inc. Supplier No. T3051L
Supplied as a component of the Monarch HMW DNA Extraction Kit for Cells & Blood, the Monarch RBC Lysis buffer is specifically designed to lyse red blood cells (erythrocytes) while leaving leukocytes intact. The separate RBC lysis step enables removal of hemoglobin to increase the purity of the blood prep. The RBC Lysis Buffer also removes nucleases to enable the subsequent purification of HMW DNA into the megabase (Mb) range in size. This product should be stored at 4°C.
  • A component of the Monarch HMW DNA Extraction Kit for Cells & Blood
  • Lyses red blood cells (erythrocytes) while leaving leukocytes intact
  • Enables removal of hemoglobin and nucleases in blood preps
  • Store Monarch RBC Lysis Buffer at 4°C