Learn More
New England Biolabs, Inc. Monarch® Spin Columns S2A and Tubes - 100 preps
SDP
Supplier:  New England Biolabs, Inc. T2037L
Encompass
The Monarch Spin Columns S1A and Tubes are a component of Monarch kits for DNA gel extraction, PCR cleanup, RNA cleanup, and also offered separately for convenience and flexibility. The columns are designed without the use of a retaining ring, helping to ensure no buffer retention and no carryover of contaminants.
  • Monarch spin columns and collection tubes designed for nucleic acid purification
  • Minimal to no buffer retention/risk of carry-over contamination
  • Low elution volume from unique column design
  • Up to 5 - 10 ug nucleic acid binding capacity, depending on application
  • Made with less plastic than conventional columns from leading suppliers
Catalog No. 50-195-9391