Learn More
New England Biolabs, Inc. Monarch® Spin RNA Cleanup Kit (10 ug) - 100 preps
SDP
Catalog No. 501959390
Encompass
The Monarch Spin RNA Cleanup Kit (10 ug) rapidly and reliably purifies up to 10 ug of concentrated, high-quality RNA (> 25 nt) from enzymatic reactions including labeling, capping, in vitro transcription (IVT) and DNase I treatment. The kit can also be applied for total RNA extraction from some sample types. This kit utilizes a bind/wash/elute workflow with minimal incubation and spin times. Our unique column design helps ensure zero buffer retention and no carryover of contaminants, enabling elution of sample in volumes as low as 6 ul. Eluted RNA is ready for use in a variety of downstream applications, including RT-PCR, RNA Library Prep for NGS and RNA labelling. The protocol can also be modified to enable the purification of smaller RNA fragments (>= 15 nts). Monarch Spin RNA Cleanup kits are also available for 50 ug and 500 ug binding capacities. Columns and buffers are also available separately for convenience
Catalog No. 50-195-9390 Supplier New England Biolabs, Inc. Supplier No. T2030L
The Monarch Spin RNA Cleanup Kit (10 ug) rapidly and reliably purifies up to 10 ug of concentrated, high-quality RNA (> 25 nt) from enzymatic reactions including labeling, capping, in vitro transcription (IVT) and DNase I treatment. The kit can also be applied for total RNA extraction from some sample types. This kit utilizes a bind/wash/elute workflow with minimal incubation and spin times. Our unique column design helps ensure zero buffer retention and no carryover of contaminants, enabling elution of sample in volumes as low as 6 ul. Eluted RNA is ready for use in a variety of downstream applications, including RT-PCR, RNA Library Prep for NGS and RNA labelling. The protocol can also be modified to enable the purification of smaller RNA fragments (>= 15 nts). Monarch Spin RNA Cleanup kits are also available for 50 ug and 500 ug binding capacities. Columns and buffers are also available separately for convenience