Learn More
New England Biolabs, Inc. Monarch® Spin Columns S2B and Tubes - 100 preps
SDP
Catalog No. NC2299112
Encompass
The Monarch Spin Columns S2B and Tubes are a component of Monarch kits for RNA cleanup, and also offered separately for convenience and flexibility. The columns are designed without the use of a retaining ring, helping to ensure no buffer retention and no carryover of contaminants.
  • Monarch spin columns and collection tubes designed for nucleic acid purification
  • Minimal to no buffer retention/risk of carry-over contamination
  • Low elution volume from unique column design
  • Included in Monarch kits, also offered separately for flexibility
  • Made with less plastic than conventional columns from leading suppliers
Catalog No. NC2299112 Supplier New England Biolabs, Inc. Supplier No. T2057L
The Monarch Spin Columns S2B and Tubes are a component of Monarch kits for RNA cleanup, and also offered separately for convenience and flexibility. The columns are designed without the use of a retaining ring, helping to ensure no buffer retention and no carryover of contaminants.
  • Monarch spin columns and collection tubes designed for nucleic acid purification
  • Minimal to no buffer retention/risk of carry-over contamination
  • Low elution volume from unique column design
  • Included in Monarch kits, also offered separately for flexibility
  • Made with less plastic than conventional columns from leading suppliers