Learn More
New England Biolabs, Inc. Monarch® Spin RNA Cleanup Kit (500 ug) - 10 preps
SDP
Catalog No. NC1657554
Encompass
The Monarch Spin RNA Cleanup Kit (500 ug) reliably purifies up to 500 ug of concentrated, high-quality RNA (>25 nt) from enzymatic reactions and in vitro transcription (IVT) reactions. The kit utilizes a bind/wash/elute workflow with minimal incubation and spin times. Our columns help ensure zero buffer retention and no carryover of contaminants, enabling elution of sample in volumes as low as 50 ul. Unwanted NTPs and short RNA fragments are removed, ensuring highly pure RNA transcripts following IVT/RNA synthesis. Eluted RNA is ready for use in a variety of downstream applications including RT-PCR, RNA Library Prep for NGS and RNA labelling. The protocol can also be modified to enable the purification of smaller RNA fragments (>= 15 nt). Monarch Spin RNA Cleanup Kits are also available for 10 ug and 50 ug binding capacities. Columns and buffers are also available separately for convenience
Catalog No. NC1657554 Supplier New England Biolabs, Inc. Supplier No. T2050S
The Monarch Spin RNA Cleanup Kit (500 ug) reliably purifies up to 500 ug of concentrated, high-quality RNA (>25 nt) from enzymatic reactions and in vitro transcription (IVT) reactions. The kit utilizes a bind/wash/elute workflow with minimal incubation and spin times. Our columns help ensure zero buffer retention and no carryover of contaminants, enabling elution of sample in volumes as low as 50 ul. Unwanted NTPs and short RNA fragments are removed, ensuring highly pure RNA transcripts following IVT/RNA synthesis. Eluted RNA is ready for use in a variety of downstream applications including RT-PCR, RNA Library Prep for NGS and RNA labelling. The protocol can also be modified to enable the purification of smaller RNA fragments (>= 15 nt). Monarch Spin RNA Cleanup Kits are also available for 10 ug and 50 ug binding capacities. Columns and buffers are also available separately for convenience