Learn More
New England Biolabs, Inc. Monarch® RNA Priming Buffer - 56 ml
SDP
Catalog No. 501527889
Encompass
Monarch RNA Priming Buffer is a component of the Monarch Total RNA Miniprep Kit. It is used to wash bound RNA to remove contaminants including salts and enzymes.
  • A component of the Monarch Total RNA Miniprep Kit
  • Used to wash bound RNA to remove contaminants including salts and enzymes
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-152-7889 Supplier New England Biolabs, Inc. Supplier No. T2013L
Monarch RNA Priming Buffer is a component of the Monarch Total RNA Miniprep Kit. It is used to wash bound RNA to remove contaminants including salts and enzymes.
  • A component of the Monarch Total RNA Miniprep Kit
  • Used to wash bound RNA to remove contaminants including salts and enzymes
  • Monarch kit components are also available separately for added convenience