Learn More
New England Biolabs, Inc. Monarch® RNA Wash Buffer - 50 ml
SDP
Catalog No. 501527890
Encompass
Monarch RNA Wash Buffer is a component of the Monarch Total RNA Miniprep Kit. It is used to wash bound RNA to remove contaminants including salts and enzymes. It is supplied as a 5X concentrate.
  • A component of the Monarch Total RNA Miniprep Kit
  • Used to wash bound RNA to remove contaminants including salts and enzymes
  • Supplied as a 5X concentrate
  • Monarch kit components are also available separately for added convenience
Catalog No. 50-152-7890 Supplier New England Biolabs, Inc. Supplier No. T2014L
Monarch RNA Wash Buffer is a component of the Monarch Total RNA Miniprep Kit. It is used to wash bound RNA to remove contaminants including salts and enzymes. It is supplied as a 5X concentrate.
  • A component of the Monarch Total RNA Miniprep Kit
  • Used to wash bound RNA to remove contaminants including salts and enzymes
  • Supplied as a 5X concentrate
  • Monarch kit components are also available separately for added convenience