Learn More
New England Biolabs, Inc. Monarch® Spin Columns S2D and Tubes - 100 units/preps
SDP
Supplier:  New England Biolabs, Inc. T1117L
Encompass
Monarch Spin Columns S2D and Tubes have been designed and assembled to deliver high performance for nucleic acid purification and concentration. They are available for purchase separately and included with Monarch Spin Nucleic Acid Purification Kits. They are designed without an O-ring, eliminating buffer retention and the risk of salt carry-over contamination, resulting in highly pure nucleic acid. The tapered column design allows for elution volumes as low as 30 ul for highly concentrated nucleic acid suitable for various downstream applications. Additionally, Monarch spin columns are designed with sustainability in mind, using less plastic than conventional columns without compromising performance. These columns are compatible for centrifugation and vacuum manifold protocols and are supplied with collection tubes.
    Includes 100 Monarch spin columns and 100 collection tubes
  • Up to 20 ug to 100 ug nucleic acid binding capacity, depending on application
Catalog No. 50-272-2967