Learn More
New England Biolabs, Inc. Monarch® StabiLyse DNA/RNA Buffer - 200 ml
SDP
Supplier:  New England Biolabs, Inc. T2111L
Encompass
The Monarch StabiLyse DNA/RNA Buffer is a dual-function reagent for nucleic acid stabilization and sample lysis. This proprietary formulation preserves the integrity of DNA and RNA in samples during storage and it also enables effective sample lysis from a wide variety of eukaryotic and prokaryotic sample types. Samples can be stored in the Monarch StabiLyse DNA/RNA Buffer at the point of harvest for short and long term without the need for immediate processing. The Monarch StabiLyse DNA/RNA Buffer is unique in that it does not require removal of this reagent or sample precipitation. The Monarch StabiLyse DNA/RNA Buffer is ideal for performing sample homogenization and mechanical disruption. The extracted nucleic acids are suitable for a variety of downstream applications including PCR, sequencing, and more
  • Dual-functioning DNA/RNA preservation & sample storage and lysis solution
Catalog No. 50-258-1802