Learn More
Medchemexpress LLC HY-N6641 5mg Medchemexpress, Monascin CAS:21516-68-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66415MG
Encompass_Preferred
Medchemexpress, HY-N6641 5mg Monascin CAS:21516-68-7 Monascin is a kind of azaphilonoid pigments extracted from Monascus pilosus-fermented rice (red-mold rice). Monascin also exhibits anti-tumor-initiating activity and anti-inflammatory activity with oral administration. Monascin inhibits the activation of NOR 1 (an NO donor). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0451