Learn More
Cayman Chemical Ankaflavin, Purity: ≥98%, CAS: 50980-32-0, 10 mg
SDP
Catalog No. 502918520
Encompass_Preferred
A microbial metabolite with diverse biological activities; induces cytotoxicity in HepG2 and A549 cancer cells (IC50 = ~15 µg/ml for both); induces cell cycle arrest at the G0 phase in HepG2 cells at 15 µg/ml; reduces NO production in LPS-stimulated RAW 264.7 macrophages at 5, 10, and 15 µg/ml; increases lipolysis activity in IBMX-, dexamethasone-, and insulin-matured 3T3-L1 adipocytes at 10 µM; reduces blood glucose and serum insulin levels in mice fed a high-fat diet at 5 mg/kg per day
Catalog No. 50-291-8520 Supplier Cayman Chemical Supplier No. 3954610 MG
A microbial metabolite with diverse biological activities; induces cytotoxicity in HepG2 and A549 cancer cells (IC50 = ~15 µg/ml for both); induces cell cycle arrest at the G0 phase in HepG2 cells at 15 µg/ml; reduces NO production in LPS-stimulated RAW 264.7 macrophages at 5, 10, and 15 µg/ml; increases lipolysis activity in IBMX-, dexamethasone-, and insulin-matured 3T3-L1 adipocytes at 10 µM; reduces blood glucose and serum insulin levels in mice fed a high-fat diet at 5 mg/kg per day