Learn More
Cayman Chemical MonascuspIloIn 2.5mg
SDP
Catalog No. 502949757
Encompass_Preferred
A fungal metabolite with anticancer activity; decreases viability of PC3 cells at 15-45 µM; has an additive effect on the reduction in viability of PC3 cells induced by irradiation at 25 µM; induces intratumor apoptosis and autophagy and reduces tumor growth in a PC3 mouse xenograft model at 40 and 120 mg/kg
Catalog No. 50-294-9757 Supplier Cayman Chemical Supplier No. 330982.5 MG
A fungal metabolite with anticancer activity; decreases viability of PC3 cells at 15-45 µM; has an additive effect on the reduction in viability of PC3 cells induced by irradiation at 25 µM; induces intratumor apoptosis and autophagy and reduces tumor growth in a PC3 mouse xenograft model at 40 and 120 mg/kg