Learn More
Medchemexpress LLC Monastrol | 329689-23-8 | MFCD00813077 | 99.9% | 292.35 g/mol | C14H16N2O3S | 100 MG
SDP
Catalog No. 5000211936
Encompass_Preferred
Monastrol is a cell-permeable small-molecule inhibitor of the mitotic kinesin Eg5, used in cell biology and mitosis research to perturb spindle dynamics and study chromosome segregation. It displays an IC50 of 14 μM against Eg5 and is provided as a high-purity research reagent for in vitro and cell-based assays.
  • Inhibits Eg5 with an IC50 of 14 μM.
  • High purity suitable for research applications.
  • Cell-permeable for use in live-cell assays.
  • Available in multiple pack sizes, including 100 MG.
  • Useful for studying spindle assembly and mitotic progression.
Catalog No. 50-002-11936 Supplier Medchemexpress LLC Supplier No. HY101071A100MG
Monastrol is a cell-permeable small-molecule inhibitor of the mitotic kinesin Eg5, used in cell biology and mitosis research to perturb spindle dynamics and study chromosome segregation. It displays an IC50 of 14 μM against Eg5 and is provided as a high-purity research reagent for in vitro and cell-based assays.
  • Inhibits Eg5 with an IC50 of 14 μM.
  • High purity suitable for research applications.
  • Cell-permeable for use in live-cell assays.
  • Available in multiple pack sizes, including 100 MG.
  • Useful for studying spindle assembly and mitotic progression.