Learn More
Abcam Monastrol, cell permeable mitosis inhibitor, 10MG
Catalog No. 502706916
Encompass_Preferred
MW 292.36 Da, Purity >99%. A potent, cell-permeable, small molecule mitosis inhibitor. It arrests cells in mitosis and specifically inhibits the motility of the mitotic kinesin Eg5, a motor protein required for mitotic spindle formation and maintenance (IC₅₀ = 14 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6916 Supplier Abcam Supplier No. AB14108710MG
MW 292.36 Da, Purity >99%. A potent, cell-permeable, small molecule mitosis inhibitor. It arrests cells in mitosis and specifically inhibits the motility of the mitotic kinesin Eg5, a motor protein required for mitotic spindle formation and maintenance (IC₅₀ = 14 μM).
The product is subject to the following: Abcam Restricted Use Statement