Learn More
TARGETMOL CHEMICALS INC Monatepil maleate 1MG
SDP
Catalog No. 502879192
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Monatepil maleate (AJ-2615 maleate) is an orally active Ca2+-channel and alpha1-adrenoceptor antagonist and non-competitive heparanoyl-coenzyme Acholesterol acyltransferase (ACAT) inhibitor with antihypertensive activity.Monatepil maleate is used in studies of hyperlipidemia and atherosclerosis. Monatepil maleate is used to study hyperlipidemia and atherosclerosis. Purity 96.28%
Catalog No. 50-287-9192 Supplier TARGETMOL CHEMICALS INC Supplier No. T258281MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Monatepil maleate (AJ-2615 maleate) is an orally active Ca2+-channel and alpha1-adrenoceptor antagonist and non-competitive heparanoyl-coenzyme Acholesterol acyltransferase (ACAT) inhibitor with antihypertensive activity.Monatepil maleate is used in studies of hyperlipidemia and atherosclerosis. Monatepil maleate is used to study hyperlipidemia and atherosclerosis. Purity 96.28%