Learn More
Cayman Chemical MonbromobImn 10mg
SDP
Catalog No. 502967560
Encompass_Preferred
A thiol-reactive fluorogenic probe; used to evaluate or quantify a variety of compounds containing reactive sulfur or thiol groups, including H2S, glutathione, proteins, and nucleotides
Catalog No. 50-296-7560 Supplier Cayman Chemical Supplier No. 1709710 MG
A thiol-reactive fluorogenic probe; used to evaluate or quantify a variety of compounds containing reactive sulfur or thiol groups, including H2S, glutathione, proteins, and nucleotides