Learn More
Cayman Chemical Monobromobimane, Purity: ≥98%, CAS: 71418-44-5, 50 mg
SDP
Catalog No. 502965901
Encompass_Preferred
A thiol-reactive fluorogenic probe; used to evaluate or quantify a variety of compounds containing reactive sulfur or thiol groups, including H2S, glutathione, proteins, and nucleotides
Catalog No. 50-296-5901 Supplier Cayman Chemical Supplier No. 1709750 MG
A thiol-reactive fluorogenic probe; used to evaluate or quantify a variety of compounds containing reactive sulfur or thiol groups, including H2S, glutathione, proteins, and nucleotides