Learn More
Morwell MD Biosciences MONELISA UMOD KIT
SDP
Supplier:  Morwell MD Biosciences M036020
Encompass
MonELISA Uromodulin Glycoprotein -Tamm-Horsfall Glycoprotein.. THP- ELISA. The MonELISA Uromodulin Glycoprotein ELISA kit is used for the quantitative determination of Uromodulin Glycoprotein -THP- in urine and serum samples. Requires MonELISA Ancillary Reagent Kit -sold separately-. Species --human/canine/mouse. Sample Type -- urine / serum
Catalog No. NC2319126