Learn More
Medchemexpress LLC Monensin methyl ester | 28636-21-7 | 99.18% | 25 MG
SDP
Catalog No. 5000342651
Encompass_Preferred
Monensin methyl ester is a neutral analog of Monensin that functions as an ion active component for Na+ selective disk electrodes. It is intended for research use only.
  • Neutral analog of Monensin
  • Functions as an ion active component for Na+ selective disk electrodes
  • Suitable for laboratory research
Catalog No. 50-003-42651 Supplier Medchemexpress LLC Supplier No. 25MGHY131142
Monensin methyl ester is a neutral analog of Monensin that functions as an ion active component for Na+ selective disk electrodes. It is intended for research use only.
  • Neutral analog of Monensin
  • Functions as an ion active component for Na+ selective disk electrodes
  • Suitable for laboratory research