Learn More
Enzo Life Sciences Monensin . sodium salt (1g). CAS: 22373-78-0
SDP
Catalog No. 502008915
Encompass_Preferred
Antibiotic that functions as Na+ ionophore. Blocks glycoprotein secretion. Inhibits proliferation of several lymphoma cell lines through cell cycle arrest and apoptosis. Blocks ceramide transport through the Golgi apparatus. Purity: ≥90%. Appearance: White to light yellow powder. Solubility: Soluble in 100% ethanol (25mg/ml), methanol (50mg/ml), chloroform or DMSO (25mg/ml). Slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8915 Supplier Enzo Life Sciences Supplier No. ALX380026G001
Antibiotic that functions as Na+ ionophore. Blocks glycoprotein secretion. Inhibits proliferation of several lymphoma cell lines through cell cycle arrest and apoptosis. Blocks ceramide transport through the Golgi apparatus. Purity: ≥90%. Appearance: White to light yellow powder. Solubility: Soluble in 100% ethanol (25mg/ml), methanol (50mg/ml), chloroform or DMSO (25mg/ml). Slightly soluble in water. Long Term Storage: -20°C.