Learn More
Medchemexpress LLC HY-14774 5mg Medchemexpress, Monepantel CAS:887148-69-8 Purity:>98%
SDP
Catalog No. 502081637
Encompass_Preferred
Medchemexpress, HY-14774 5mg Monepantel CAS:887148-69-8 Monepantel is organic anthelmintic, and acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1637 Supplier Medchemexpress LLC Supplier No. HY147745MG
Medchemexpress, HY-14774 5mg Monepantel CAS:887148-69-8 Monepantel is organic anthelmintic, and acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.