Learn More
Medchemexpress LLC Monepantel (Standard) | 887148-69-8 | 99.5% | 473.39 | 1 MG
SDP
Catalog No. 5000362223
Encompass_Preferred
Monepantel (Standard) is an analytical standard of Monepantel, intended for research and analytical applications. It functions as an organic anthelmintic and acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits. This product is suitable for qualitative, quantitative, and methodological research experiments utilizing techniques such as HPLC, GC, and MS.
  • Serves as a reference standard for assays
  • Primarily intended for research and analytical applications
  • Acts as a positive allosteric modulator of nematode-specific nicotinic acetylcholine receptor (nAChR) subunits
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for use with techniques including HPLC, GC, and MS
Catalog No. 50-003-62223 Supplier Medchemexpress LLC Supplier No. HY14774R1MG
Monepantel (Standard) is an analytical standard of Monepantel, intended for research and analytical applications. It functions as an organic anthelmintic and acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits. This product is suitable for qualitative, quantitative, and methodological research experiments utilizing techniques such as HPLC, GC, and MS.
  • Serves as a reference standard for assays
  • Primarily intended for research and analytical applications
  • Acts as a positive allosteric modulator of nematode-specific nicotinic acetylcholine receptor (nAChR) subunits
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for use with techniques including HPLC, GC, and MS