Learn More
Medchemexpress LLC HY-101905 1mg Medchemexpress, Moniliformin (sodium salt) CAS:71376-34-6 Purity:>98%
SDP
Catalog No. 501932227
Encompass_Preferred
Medchemexpress, HY-101905 1mg Moniliformin (sodium salt) CAS:71376-34-6 Moniliformin sodium salt is a potent, water-soluble mycotoxin isolate from Fusarium moniliforme. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2227 Supplier Medchemexpress LLC Supplier No. HY1019051MG
Medchemexpress, HY-101905 1mg Moniliformin (sodium salt) CAS:71376-34-6 Moniliformin sodium salt is a potent, water-soluble mycotoxin isolate from Fusarium moniliforme. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.