Learn More
Enzo Life Sciences Moniliformin. sodium salt (1mg). CAS: 71376-34-6
SDP
Supplier:  Enzo Life Sciences ALX630111M001
Encompass_Preferred
Mycotoxin. Alternative name: 1-Hydroxycyclobut-1-ene-3,4-dione. Purity: ≥98% (TLC). Appearance: Light yellow powder. Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9636