Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moniliformin sodium salt | 71376-34-6 | MFCD01714659 | 99.6% | 120.04 g/mol | C4HNaO3 | 1 ML
SDP

Catalog No. 5000249157
Encompass_Preferred

Moniliformin sodium salt is the sodium salt form of moniliformin, a mycotoxin isolated from Fusarium moniliforme. Supplied for research use, this light yellow solid is characterized by a reported high purity and provided in small research quantities either as a solid or as a DMSO solution. Product documentation includes solubility and storage guidance to support preparation and handling in laboratory studies.

  • High purity: 99.62% as reported by certificate of analysis.
  • Available as solid or DMSO solution for flexible experimental use.
  • Soluble in water and DMSO with specified concentrations for formulation.
  • Light yellow solid appearance suitable for analytical and in vitro work.
  • Recommended storage: solid at 4°C; in solvent, -80°C for long-term storage.

Catalog No. 50-002-49157 Supplier Medchemexpress LLC Supplier No. HY10190510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moniliformin sodium salt is the sodium salt form of moniliformin, a mycotoxin isolated from Fusarium moniliforme. Supplied for research use, this light yellow solid is characterized by a reported high purity and provided in small research quantities either as a solid or as a DMSO solution. Product documentation includes solubility and storage guidance to support preparation and handling in laboratory studies.

  • High purity: 99.62% as reported by certificate of analysis.
  • Available as solid or DMSO solution for flexible experimental use.
  • Soluble in water and DMSO with specified concentrations for formulation.
  • Light yellow solid appearance suitable for analytical and in vitro work.
  • Recommended storage: solid at 4°C; in solvent, -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.