Learn More
Molecular Devices Molecular Devices Genomic Filter Slide, 260/280/320 nm, 10 nm Bandwidth
Catalog No. 5000014688
Encompass_Preferred
The Molecular Devices Genomic Filter Slide is a multi-filter slide for filter-based microplate readers, built for genomic absorbance measurements and nucleic acid quantitation. It contains narrow-band UV filters that enable accurate 260/280 ratio assessments for nucleic acid purity.
  • Contains filters centered at 260 nm, 280 nm, and 320 nm.
  • 10 nm bandwidth for each filter.
  • Designed for genomic absorbance and nucleic acid quantitation.
  • Fits standard filter-slide mounts on compatible microplate readers.
  • Enables reliable measurement of nucleic acid purity via 260/280 ratios.
Catalog No. 50-000-14688 Supplier Molecular Devices Supplier No. 5314902
The Molecular Devices Genomic Filter Slide is a multi-filter slide for filter-based microplate readers, built for genomic absorbance measurements and nucleic acid quantitation. It contains narrow-band UV filters that enable accurate 260/280 ratio assessments for nucleic acid purity.
  • Contains filters centered at 260 nm, 280 nm, and 320 nm.
  • 10 nm bandwidth for each filter.
  • Designed for genomic absorbance and nucleic acid quantitation.
  • Fits standard filter-slide mounts on compatible microplate readers.
  • Enables reliable measurement of nucleic acid purity via 260/280 ratios.