Learn More
Med Vet International Home Automatic Digital Blood Pressure Monitor Drive™ Medium Cuff Nylon Cuff 22 - 36 cm Desk Model, BP3400, Each
Catalog No. 503045540
Encompass_Preferred
A one-touch operation provides for convenient usage The large, easy-to-read LCD display makes it easy to obtain results This adult blood pressure monitor measures pulse It alerts users to the presence of an irregular heartbeat Drive Medical automatic deluxe blood pressure monitor stores up to 120 readings for a total of two users It requires four AA batteries An auto-off feature saves battery life The upper arm digital blood pressure monitor is a medium cuff
Catalog No. 50-304-5540 Supplier Med Vet International Supplier No. 1055467EA
A one-touch operation provides for convenient usage The large, easy-to-read LCD display makes it easy to obtain results This adult blood pressure monitor measures pulse It alerts users to the presence of an irregular heartbeat Drive Medical automatic deluxe blood pressure monitor stores up to 120 readings for a total of two users It requires four AA batteries An auto-off feature saves battery life The upper arm digital blood pressure monitor is a medium cuff