Learn More
Med Vet International Home Automatic Digital Blood Pressure Monitor MABIS One Size Fits Most Cuff Nylon Cuff 13 - 21 cm Wrist, 4-810-001, Each
Catalog No. 503045552
Encompass_Preferred
The battery-powered Mabis digital wrist blood pressure monitor is compact, cord-free and portable A backlit LCD screen shows large, easy-to-read numbers Readings of pulse and blood pressure are clinic-accurate for reliable performance Irregular heartbeat detection alerts users in real time A visual guide shows where BP falls on a low-to-high scale The Mabis blood pressure monitor stores up to 120 readings for one user or 60 each for two users, providing reference data The unit establishes a baseline by averaging the last three readings
Catalog No. 50-304-5552 Supplier Med Vet International Supplier No. 1012669EA
The battery-powered Mabis digital wrist blood pressure monitor is compact, cord-free and portable A backlit LCD screen shows large, easy-to-read numbers Readings of pulse and blood pressure are clinic-accurate for reliable performance Irregular heartbeat detection alerts users in real time A visual guide shows where BP falls on a low-to-high scale The Mabis blood pressure monitor stores up to 120 readings for one user or 60 each for two users, providing reference data The unit establishes a baseline by averaging the last three readings