Learn More
Med Vet International Home Automatic Digital Blood Pressure Monitor Welch Allyn™ Home™ Wide Range Nylon Cuff 22 - 42 cm Desk Model, H-BP100SBP, Each
Catalog No. 503045555
Encompass_Preferred
Captures blood pressure on inflation for quick and comfortable readings Irregular heartbeat indicator Motion indicator Free app enables patients to engage in their progress toward blood pressure goals and automatically send readings to their physician for review Free Clinical Portal allows physicians to securely review patient compliance and progress Powered by four AA alkaline batteries (included) AC adapter available Welch Allyn Home Blood Pressure Monitor with SureBP (1700 Series) with D-Ring Standard Wide Cuff (22-42cm), 4 Alkaline AA batteries Extra Small Cuffs (15-24cm)and Extra Large Blood Pressure Cuffs (40-54cm) are available for purchase
Catalog No. 50-304-5555 Supplier Med Vet International Supplier No. 1041130EA
 Restricted Item
Add to Cart
Add to Cart
Captures blood pressure on inflation for quick and comfortable readings Irregular heartbeat indicator Motion indicator Free app enables patients to engage in their progress toward blood pressure goals and automatically send readings to their physician for review Free Clinical Portal allows physicians to securely review patient compliance and progress Powered by four AA alkaline batteries (included) AC adapter available Welch Allyn Home Blood Pressure Monitor with SureBP (1700 Series) with D-Ring Standard Wide Cuff (22-42cm), 4 Alkaline AA batteries Extra Small Cuffs (15-24cm)and Extra Large Blood Pressure Cuffs (40-54cm) are available for purchase