Learn More
Med Vet International Home Automatic Digital Blood Pressure Monitor Smartheart Adult Cuff Nylon Cuff 22 to 42 cm Desk Model, 01-509, 12/Case
Catalog No. 503045538
Encompass_Preferred
Cuff fits arm size: 8.6” – 16.5” (22–42cm) Clinically accurate readings Fully automatic inflation and deflation Systolic, diastolic and pulse results 2-person memory bank holds 500-readings, 250 per user Hypertension Indicator Irregular Heartbeat Detection Includes: Monitor with an attached nylon arm cuff, storage case, DC wall adapter, internal rechargeable battery, detailed English/Spanish instructions, quick start guide, FAQ pamphlet and blood pressure charting log
Catalog No. 50-304-5538 Supplier Med Vet International Supplier No. 1226102CS
Cuff fits arm size: 8.6” – 16.5” (22–42cm) Clinically accurate readings Fully automatic inflation and deflation Systolic, diastolic and pulse results 2-person memory bank holds 500-readings, 250 per user Hypertension Indicator Irregular Heartbeat Detection Includes: Monitor with an attached nylon arm cuff, storage case, DC wall adapter, internal rechargeable battery, detailed English/Spanish instructions, quick start guide, FAQ pamphlet and blood pressure charting log