Learn More
Cole-Parmer Oakton ECCONCP0550 COND 550 Conductivity Monitor, 110/220VAC
Catalog No. 501961737 Shop All Cole-Parmer Products
Encompass_Preferred
The CON 550 is a small, economical monitor used to measure conductivity and temperature values in applications such as water treatment and monitoring, plating, chemical processing, or food processing. The monitor has user-friendly features like menu-driven programs and calibrations that are easily performed with the push button key pad. All setup and calibration parameters are retained in nonvolatile memory to prevent data loss during power outages. Measurements can be compensated for temperature variations automatically or manually. Independent calibration and process temperatures can be set when manual temperature compensation is used.
Catalog No. 50-196-1737 Supplier Cole-Parmer Supplier No. 1950530
The CON 550 is a small, economical monitor used to measure conductivity and temperature values in applications such as water treatment and monitoring, plating, chemical processing, or food processing. The monitor has user-friendly features like menu-driven programs and calibrations that are easily performed with the push button key pad. All setup and calibration parameters are retained in nonvolatile memory to prevent data loss during power outages. Measurements can be compensated for temperature variations automatically or manually. Independent calibration and process temperatures can be set when manual temperature compensation is used.