Learn More
Electron Microscopy Sciences Anti-Vibration Platform Monitor Stand (Needs Guard)
SDP
Catalog No. 5034462
Encompass_Preferred
Supports monitors weighing up to 30kg. Features 360 degree rotation of the arm as well as height adjustment. It mounts to the platform guard.
Catalog No. 50-344-62 Supplier Electron Microscopy Sciences Supplier No. 6540
Supports monitors weighing up to 30kg. Features 360 degree rotation of the arm as well as height adjustment. It mounts to the platform guard.