Learn More
Essendant Refrigerator/Freezer Monitoring Thermometer, -20F to 80F
Catalog No. 2679320
Encompass_Preferred
Prevent food spoilage by accurately monitoring your refrigerator and freezer interior temperatures. Large Celsius and Fahrenheit scales enable at-a-glance reads. Shatter-resistant lens minimizes the potential for breakage. Thermometer may be mounted horizontally or vertically.
Catalog No. 26-793-20 Supplier Essendant Supplier No. PELR80DC
Prevent food spoilage by accurately monitoring your refrigerator and freezer interior temperatures. Large Celsius and Fahrenheit scales enable at-a-glance reads. Shatter-resistant lens minimizes the potential for breakage. Thermometer may be mounted horizontally or vertically.