Learn More
Invitrogen™ Rhesus Monkey ANGPT2 ELISA Kit
Description
Monkey ANGPT2 quantitates monkey ANGPT2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey ANGPT2.
Specifications
Specifications
| Assay Range | 0.12-30 ng/mL |
| Assay Sensitivity | 0.12 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Monkey |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | AGPT2; Ang2; Ang-2; angiopoietin 2; angiopoietin-2; angiopoietin-2a; angiopoietin-2B; ANGPT2; endothelial growth factor; Tie2-ligandAGPT2; Ang2; Ang-2; angiopoietin 2; angiopoietin-2; angiopoietin-2a; angiopoietin-2B; ANGPT2; endothelial growth factor; Tie2-ligand; |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.