Learn More
Life Diagnostics Inc Monkey Anti-PEG IgG ELISA, 1000-fold initial sample dilution, 450 nm detection, 2 hours 5 minutes assay time, solid phase ELISA kit
SDP
Supplier:  Life Diagnostics Inc PEGG3N
Encompass_Preferred
Monkey Anti-PEG IgG ELISA, 1000-fold initial sample dilution, 450 nm detection, 2 hours 5 minutes assay time, solid phase ELISA kit
Catalog No. NC1463865