Learn More
American Research Products Inc Monkey apolipoprotein A1 Apo
Catalog No. 5000330099
Encompass
Monkey apolipoprotein A1 (Apo-A1) ELISA Kit from CUSABIO Detection Range 0 7 ng/mL-500 ng/mL Sensitivity 0 7 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-30099 Supplier American Research Products Inc Supplier No. CSBE11041MO
Monkey apolipoprotein A1 (Apo-A1) ELISA Kit from CUSABIO Detection Range 0 7 ng/mL-500 ng/mL Sensitivity 0 7 ng/mL Sample serum plasma Method Sandwich