Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monkey BDNF ELISA Kit

Catalog No. EP20RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EP20RBX5 5 x 96 Tests
EP20RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EP20RBX5 Supplier Invitrogen™ Supplier No. EP20RBX5
Add to Cart
Add to Cart

ELISA

Monkey BDNF quantitates monkey BDNF in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey BDNF.

BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF
Assay Range 2.5-500 pg/mL
Assay Sensitivity 2.5 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Monkey
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNFabrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF;
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target BDNF
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.